Assay Details
| Gene Symbol: 5031439G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033273.2 | NP_001028445.1 | 3 - 4 | 639 | 66 | |
| GenBank mRNA | AK138575.1 | - | 2 - 3 | 624 | 66 | |
| AK149782.1 | - | 3 - 4 | 640 | 66 | ||
| AK170532.1 | - | 3 - 4 | 672 | 66 | ||
| AK173065.1 | - | 3 - 4 | 397 | 66 | ||
More Information
Updated Mapping Information
Set Membership: | 
		   
					
						
							 | 
				

5031439G07Rik
Genomic Map