Assay Details
| Gene Symbol: 5031439G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033273.2 | NP_001028445.1 | 7 - 8 | 1154 | 97 | |
| GenBank mRNA | AK149782.1 | - | 7 - 8 | 1155 | 97 | |
| AK170532.1 | - | 7 - 8 | 1187 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

5031439G07Rik
Genomic Map