Assay Details
Gene Symbol: Tymp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138302.1 | NP_612175.1 | 1 - 2 | 64 | 128 | |
GenBank mRNA | AB060274.1 | - | 1 - 2 | 64 | 128 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB060274.1, NM_138302.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
