Assay Details
| Gene Symbol: Zfp449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030139.4 | NP_084415.1 | 4 - 5 | 807 | 159 | |
| GenBank mRNA | AK036596.1 | - | 4 - 5 | 808 | 159 | |
| AK142888.1 | - | 4 - 5 | 812 | 159 | ||
| BC082594.1 | - | 3 - 4 | 697 | 30642886 | 159 | |
| BC145936.1 | - | 3 - 4 | 697 | 40131239 | 159 | |
| BC145938.1 | - | 3 - 4 | 697 | 40131241 | 159 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32395553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32395552 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | BC145938.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | BC082594.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | BC145936.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | AK036596.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | AK142888.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | BC145938.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | BC082594.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | BC145936.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | AK036596.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263844 | AK142888.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920607 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304882 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394194 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696802 | NM_146291.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132843 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127139 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41958 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165074 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | BC145938.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | BC082594.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | BC145936.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | AK036596.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | AK142888.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | BC145938.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | BC082594.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | BC145936.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | AK036596.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263844 | AK142888.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P1200510 | BC082594.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P1200510 | NM_030139.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | BC145938.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | BC082594.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | BC145936.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | AK036596.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | AK142888.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | BC145938.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | BC082594.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | BC145936.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | AK036596.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263844 | AK142888.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32395511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32395510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32395503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32395502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32395512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32395504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32395509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32395506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32395505 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165074 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09298954 | - | - |
| Whole Mouse Genome G4122A | A_52_P1200510 | BC082594.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1200510 | NM_030139.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | BC145938.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | BC082594.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | BC145936.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | AK036596.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | AK142888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | BC145938.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | BC082594.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | BC145936.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | AK036596.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263844 | AK142888.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122907 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09298954 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165074 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15218364 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15218366 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15218374 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32395553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32395552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32395540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15052207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15044379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15044380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14832542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15044375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32395527 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10599690 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599687 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599689 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599688 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599691 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439945_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10599686 | - | - |
| Mouse Genome 430 2.0 Array | 1439945_at | - | - |
| Mouse Genome 430 2.0 Array | 1439945_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78619_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78619_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00003366.mm.1 | NM_030139.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00003366.mm.1 | NM_030139.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00003366.mm.1 | TC0X00000515.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00003367.mm.1 | NM_030139.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00003367.mm.1 | NM_030139.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00003367.mm.1 | TC0X00000515.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439945_at | - | - |
| Mouse Expression Set 430 | 1439945_PM_at | - | - |
| Murine Genome U74v2 Set | 134597_at | - | - |
| Murine Genome U74v2 Set | 134597_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439945_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439945_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6590154 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6590154 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590154 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226584 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226584 | - | - |
| mouse-6 v1.1 (Illumina) | scl54925.5_509-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590154 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54925.5_509-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590154 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226584 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226584 | - | - |

Zfp449
Genomic Map