Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300017600.mm.1 |
NM_001005868.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017600.mm.1 |
NM_001289473.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017600.mm.1 |
NM_001289475.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017600.mm.1 |
TC1300002588.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300017605.mm.1 |
NM_001005868.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017605.mm.1 |
NM_001289475.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017605.mm.1 |
TC1300002588.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300017599.mm.1 |
NM_001005868.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017599.mm.1 |
NM_001289473.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017599.mm.1 |
NM_001289475.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017599.mm.1 |
TC1300002588.mm.1 |
MTA TC |