Assay Details
Gene Symbol: Gm9530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029591.1 | - | 4 - 5 | 2546 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
calcium channel, voltage-dependent, R type, alpha 1E subunit |
Cacna1e |
A430040I15, BII, Cach6, Cacnl1a6, Cav2.3, Cchra1, alpha1E |
-180.4kb | Sense | Chr. 1 - 154390806 - 154884782 on NCBI Build GRCm38 |
Downstream | immediate early response 5 | Ier5 | 30.5kb | Sense | Chr. 1 - 155096367 - 155099636 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
