Assay Details
Gene Symbol: Gm9530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029591.1 | - | 6 - 7 | 2872 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
calcium channel, voltage-dependent, R type, alpha 1E subunit |
Cacna1e |
A430040I15, BII, Cach6, Cacnl1a6, Cav2.3, Cchra1, alpha1E |
-178.7kb | Sense | Chr. 1 - 154390806 - 154884782 on NCBI Build GRCm38 |
Downstream | immediate early response 5 | Ier5 | 31.3kb | Sense | Chr. 1 - 155096367 - 155099636 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P138363 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781199 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145825 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63781 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | AK013944.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | AK145977.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | AK146378.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | AK151750.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | AK152689.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | BC003233.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | BC023244.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | BC050882.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | BC082274.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | NM_016849.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | U75839.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925384 | U75840.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150821 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605869 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502854 | AK035829.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848013 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80485 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04315400 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04315430 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138363 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04315430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735582 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735685 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00665295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735652 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735548 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735697 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735677 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735574 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00665301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735694 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735667 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735682 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00515081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00767174 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00528513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735579 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00665284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01045201 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735705 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735687 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735580 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735666 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00981295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00749694 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735551 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735622 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01033362 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00322297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00800928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735683 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735654 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00241613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735668 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01045202 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00322280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00665292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735638 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735553 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00993525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735673 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00285529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735700 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00767165 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735520 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735729 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01033361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735699 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735672 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00322282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00994084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01045200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735552 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735679 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00269406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00767163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735578 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735576 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00745992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735643 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00689383 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735522 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735651 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735721 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735577 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735690 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00285530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00822117 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20735663 | - | - |
