Assay Details
| Gene Symbol: Il13ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080147.1 | - | 5 - 6 | 697 | 99 | |
| AK082889.1 | - | 5 - 6 | 692 | 99 | ||
| AK089984.1 | - | 5 - 6 | 728 | 99 | ||
| AK143292.1 | - | 5 - 6 | 697 | 99 | ||
| AK154675.1 | - | 5 - 6 | 752 | 99 | ||
| BC021472.1 | - | 1 - 2 | 151 | 99 | ||
| BC052425.1 | - | 5 - 6 | 720 | 6835500 | 99 | |
| BC059939.1 | - | 5 - 6 | 688 | 6477224 | 99 | |
| S80963.1 | - | 5 - 6 | 725 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il13ra1
Genomic Map