Assay Details
| Gene Symbol: Hsd17b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008291.3 | NP_032317.2 | 5 - 6 | 516 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd17b3| Gene Symbol: Hsd17b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008291.3 | NP_032317.2 | 5 - 6 | 516 | 97 | |
Set Membership: |
|