Assay Details
| Gene Symbol: Dnajc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019795.4 | NP_062769.2 | 6 - 7 | 1163 | 62 | |
| GenBank mRNA | AB028861.1 | - | 6 - 7 | 611 | 62 | |
| AK011875.1 | - | 6 - 7 | 602 | 62 | ||
| AK145506.1 | - | 6 - 7 | 625 | 62 | ||
| AK145739.1 | - | 6 - 7 | 625 | 62 | ||
| AK166872.1 | - | 6 - 7 | 625 | 62 | ||
| AY323828.1 | - | 6 - 7 | 599 | 62 | ||
| BC023681.1 | - | 6 - 7 | 807 | 5343384 | 62 | |
| BC055729.1 | - | 6 - 7 | 1162 | 6414499 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc7
Genomic Map