Assay Details
| Gene Symbol: Dhx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030150.2 | NP_084426.2 | 6 - 7 | 909 | 66 | |
| GenBank mRNA | AF316999.1 | - | 5 - 6 | 808 | 66 | |
| AK153342.1 | - | 6 - 7 | 838 | 66 | ||
| BC029209.1 | - | 5 - 6 | 819 | 2651254 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx58
Genomic Map