Assay Details
| Gene Symbol: Dhx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030150.2 | NP_084426.2 | 7 - 8 | 1094 | 80 | |
| GenBank mRNA | AF316999.1 | - | 6 - 7 | 993 | 80 | |
| AK153342.1 | - | 7 - 8 | 1023 | 80 | ||
| BC029209.1 | - | 6 - 7 | 1004 | 2651254 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx58
Genomic Map