Assay Details
Gene Symbol: Nt5c3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102650.1 | NP_001096120.1 | 8 - 9 | 805 | 70 | |
GenBank mRNA | AK148222.1 | - | 8 - 9 | 781 | 70 | |
AK162770.1 | - | 7 - 8 | 883 | 70 | ||
BC015307.1 | - | 7 - 8 | 760 | 4239117 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28068129 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28068130 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28068127 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156462 | AK148222.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156462 | AK162770.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156462 | AK166393.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156462 | AK211190.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156462 | BC015307.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156464 | AK148222.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156464 | AK162770.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156464 | BC015307.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156464 | NM_001102650.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156464 | NM_026561.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702599 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77379 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635878 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363770 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299388 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313869 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830045 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651953 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139453 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115876 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147187 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107241 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178653 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156464 | AK148222.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156464 | AK162770.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156464 | BC015307.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156464 | NM_001102650.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156464 | NM_026561.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156462 | AK148222.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156462 | AK162770.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156462 | AK166393.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156462 | AK211190.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156462 | BC015307.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068128 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068133 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068134 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068114 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178653 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07463784 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107241 | - | - |
Whole Mouse Genome G4122A | A_52_P639997 | AK148222.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P639997 | AK162770.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P639997 | BC015307.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P639997 | BY047327.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P639997 | NM_001102650.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130191 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107241 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178653 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09133635 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09314589 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09314592 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09133636 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09133630 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28068129 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28068127 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28068130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09068141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09260912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09244065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09589156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09068140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09068132 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28068127 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10391135 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391136 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391137 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391138 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391131 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381129 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391139 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391134 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391133 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391132 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451180_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449633_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1430559_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10381122 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10391130 | - | - |
Mouse Genome 430 2.0 Array | 1451180_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1430559_at | - | - |
Mouse Genome 430 2.0 Array | 1451180_a_at | - | - |
Mouse Genome 430 2.0 Array | 1449633_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1430559_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449633_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68106_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68106_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100034647.mm.1 | NM_001102650.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034647.mm.1 | NM_001102650.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034647.mm.1 | TC1100003771.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100034648.mm.1 | NM_001102650.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034648.mm.1 | NM_001102650.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034648.mm.1 | TC1100003771.mm.1 | MTA TC |
Mouse Expression Set 430 | 1451180_a_at | - | - |
Mouse Expression Set 430 | 1430559_at | - | - |
Mouse Expression Set 430 | 1451180_PM_a_at | - | - |
Mouse Expression Set 430 | 1430559_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1430559_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1430559_at | - | - |
Mouse Genome 430A 2.0 Array | 1451180_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451180_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451180_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449633_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449633_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430559_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430559_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451180_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 755631 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5290369 | - | - |
MouseWG-6 v2 Expression BeadChip | 6370685 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5290369 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6370685 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2850427 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005290369 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006370685 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002850427 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2666971 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2708357 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245842 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1245842 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2666971 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2708357 | - | - |
mouse-6 v1.1 (Illumina) | scl0001342.1_29-S | - | - |
mouse-6 v1.1 (Illumina) | scl0001518.1_3-S | - | - |
mouse-6 v1.1 (Illumina) | scl39559.14.1_100-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5290369 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6370685 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2850427 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001518.1_3-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39559.14.1_100-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001342.1_29-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370685 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850427 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5290369 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2708357 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1245842 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245842 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2708357 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001342.1_29-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001518.1_3-S | - | - |
