Assay Details
Gene Symbol: Hap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010404.3 | NP_034534.1 | 5 - 6 | 927 | 77 | |
NM_177981.2 | NP_817090.1 | 5 - 6 | 927 | 77 | ||
GenBank mRNA | AJ000262.1 | - | 5 - 6 | 873 | 77 | |
AJ002271.1 | - | 5 - 6 | 873 | 77 | ||
AK138436.1 | - | 875 | 77 | |||
BC034089.1 | - | 5 - 6 | 865 | 4481735 | 77 | |
BC053043.1 | - | 5 - 6 | 914 | 5694628 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
