Assay Details
| Gene Symbol: Trappc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC106114.1 | - | 5 - 6 | 482 | 4168174 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc6a| Gene Symbol: Trappc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC106114.1 | - | 5 - 6 | 482 | 4168174 | 85 |
Set Membership: |
|