Assay Details
| Gene Symbol: Hnf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291268.1 | NP_001278197.1 | 6 - 7 | 1446 | 82 | |
| NM_001291269.1 | NP_001278198.1 | 6 - 7 | 1155 | 82 | ||
| NM_009330.3 | NP_033356.2 | 6 - 7 | 1524 | 82 | ||
| GenBank mRNA | AB008174.1 | - | 6 - 7 | 1344 | 82 | |
| AB008175.1 | - | 6 - 7 | 1266 | 82 | ||
| AB008176.1 | - | 6 - 7 | 1344 | 82 | ||
| AB008177.1 | - | 6 - 7 | 1266 | 82 | ||
| AB052659.1 | - | 6 - 7 | 1344 | 82 | ||
| AK004837.2 | - | 6 - 7 | 1525 | 82 | ||
| AK019258.1 | - | 6 - 7 | 1078 | 82 | ||
| BC025189.1 | - | 6 - 7 | 1155 | 5044299 | 82 | |
| X55842.1 | - | 6 - 7 | 1347 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hnf1b
Genomic Map