Assay Details
| Gene Symbol: Ddx52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030096.2 | NP_084372.2 | 10 - 11 | 1445 | 109 | |
| GenBank mRNA | AK080767.1 | - | 10 - 11 | 1383 | 109 | |
| AK133545.1 | - | 10 - 11 | 1416 | 109 | ||
| AK141200.1 | - | 10 - 11 | 110 | |||
| AK166789.1 | - | 10 - 11 | 1371 | 109 | ||
| BC029094.1 | - | 10 - 11 | 1357 | 3662249 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx52
Genomic Map