Assay Details
| Gene Symbol: Ddx52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030096.2 | NP_084372.2 | 12 - 13 | 1674 | 113 | |
| GenBank mRNA | AK080767.1 | - | 12 - 13 | 1612 | 113 | |
| AK133545.1 | - | 12 - 13 | 1645 | 113 | ||
| AK141200.1 | - | 12 - 13 | 1649 | 113 | ||
| AK166789.1 | - | 12 - 13 | 1600 | 113 | ||
| BC029094.1 | - | 12 - 13 | 1586 | 3662249 | 113 | |
More Information
Set Membership: |
|

Ddx52
Genomic Map