Assay Details
| Gene Symbol: Ddx52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030096.2 | NP_084372.2 | 2 - 3 | 369 | 81 | |
| GenBank mRNA | AK133545.1 | - | 2 - 3 | 340 | 81 | |
| AK141200.1 | - | 2 - 3 | 343 | 81 | ||
| AK166789.1 | - | 2 - 3 | 295 | 81 | ||
| CJ142321.1 | - | 2 - 3 | 370 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx52
Genomic Map