Assay Details
Gene Symbol: Ddx52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030096.2 | NP_084372.2 | 6 - 7 | 954 | 114 | |
GenBank mRNA | AK080767.1 | - | 6 - 7 | 892 | 114 | |
AK133545.1 | - | 6 - 7 | 925 | 114 | ||
AK141200.1 | - | 6 - 7 | 928 | 114 | ||
AK166789.1 | - | 6 - 7 | 880 | 114 | ||
BC029094.1 | - | 6 - 7 | 866 | 3662249 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
