Assay Details
| Gene Symbol: Ccl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009139.3 | NP_033165.1 | 3 - 4 | 383 | 102 | |
| GenBank mRNA | AF128188.1 | - | 3 - 4 | 279 | 102 | |
| AF128189.1 | - | 3 - 4 | 279 | 102 | ||
| AF128190.1 | - | 3 - 4 | 279 | 102 | ||
| AF128191.1 | - | 3 - 4 | 279 | 102 | ||
| AF128192.1 | - | 3 - 4 | 279 | 102 | ||
| AK002697.1 | - | 3 - 4 | 286 | 102 | ||
| AK008547.1 | - | 3 - 4 | 287 | 102 | ||
| AK150336.1 | - | 3 - 4 | 286 | 102 | ||
| AK154553.1 | - | 3 - 4 | 287 | 102 | ||
| M58004.1 | - | 3 - 4 | 286 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | AK150336.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | AK154553.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | M58004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | NM_009139.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | AK002697.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | AK150336.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | AK154553.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | M58004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | NM_009139.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460954 | AK002697.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 577321 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103866 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111656 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | AK150336.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | AK154553.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | M58004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | NM_009139.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | AK002697.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | AK150336.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | AK154553.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | M58004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | NM_009139.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460954 | AK002697.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | AK150336.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | AK154553.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | M58004.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | NM_009139.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | AK002697.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | AK150336.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | AK154553.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | M58004.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | NM_009139.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460954 | AK002697.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972390 | - | - |
| Whole Mouse Genome G4122A | A_51_P460954 | AK150336.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460954 | AK154553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460954 | M58004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460954 | NM_009139.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460954 | AK002697.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460954 | AK150336.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460954 | AK154553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460954 | M58004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460954 | NM_009139.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460954 | AK002697.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121697 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09454622 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09454625 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09454623 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09454620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09454630 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09454616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09021751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09343403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09186466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09085483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09021764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09021742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09343399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09450968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09085486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09021740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972379 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10389228 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389224 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389225 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389223 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417266_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420249_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10389222 | - | - |
| Mouse Genome 430 2.0 Array | 1420249_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1417266_at | - | - |
| Mouse Genome 430 2.0 Array | 1420249_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1417266_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20305_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20305_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100031138.mm.1 | NM_009139.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031138.mm.1 | NM_009139.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031138.mm.1 | TC1100003399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031136.mm.1 | NM_009139.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031136.mm.1 | NM_009139.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031136.mm.1 | TC1100003399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031143.mm.1 | NM_009139.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031143.mm.1 | NM_009139.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031143.mm.1 | TC1100003399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031141.mm.1 | NM_009139.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031141.mm.1 | NM_009139.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031141.mm.1 | TC1100003399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031145.mm.1 | NM_009139.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031145.mm.1 | NM_009139.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031145.mm.1 | TC1100003399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031139.mm.1 | NM_009139.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031139.mm.1 | NM_009139.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031139.mm.1 | TC1100003399.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417266_PM_at | - | - |
| Mouse Expression Set 430 | 1420249_PM_s_at | - | - |
| Mouse Expression Set 430 | 1420249_s_at | - | - |
| Mouse Expression Set 430 | 1417266_at | - | - |
| Murine Genome U74v2 Set | 92849_at | - | - |
| Murine Genome U74v2 Set | 92849_PM_at | - | - |
| Murine Genome U74v2 Set | 167582_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167582_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417266_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417266_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420249_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420249_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417266_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417266_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420249_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420249_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6660390 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660390 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244358 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244358 | - | - |
| mouse-6 v1.1 (Illumina) | scl39817.6_54-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660390 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39817.6_54-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660390 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2694179 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694179 | - | - |

Ccl6
Genomic Map