Assay Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172562.3 | NP_766150.1 | 10 - 11 | 824 | 132 | |
| GenBank mRNA | AK157938.1 | - | 10 - 11 | 817 | 132 | |
| BC005610.1 | - | 7 - 8 | 551 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tada2a
Genomic Map