Assay Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142262.1 | - | 1 - 2 | 70 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK142262.1 transcripts.
Set Membership: |
|

Tada2a
Genomic Map