Assay Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172562.3 | NP_766150.1 | 4 - 5 | 309 | 119 | |
| GenBank mRNA | AK142262.1 | - | 4 - 5 | 341 | 119 | |
| AK157938.1 | - | 4 - 5 | 302 | 119 | ||
| BC038821.1 | - | 4 - 5 | 312 | 3990023 | 119 | |
More Information
Set Membership: |
|

Tada2a
Genomic Map