Assay Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172562.3 | NP_766150.1 | 8 - 9 | 712 | 142 | |
| GenBank mRNA | BC005610.1 | - | 5 - 6 | 439 | 142 | |
| BC025448.1 | - | 3 - 4 | 204 | 142 | ||
| BC038821.1 | - | 8 - 9 | 715 | 3990023 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tada2a
Genomic Map