Assay Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172562.3 | NP_766150.1 | 9 - 10 | 785 | 116 | |
| GenBank mRNA | AK157938.1 | - | 9 - 10 | 778 | 116 | |
| BC005610.1 | - | 6 - 7 | 512 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tada2a
Genomic Map