Assay Details
| Gene Symbol: Prss41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027644.1 | NP_081920.1 | 4 - 5 | 312 | 88 | |
| GenBank mRNA | AB049453.1 | - | 4 - 5 | 312 | 88 | |
| AK016509.1 | - | 4 - 5 | 290 | 88 | ||
| AK049297.1 | - | 4 - 5 | 329 | 88 | ||
| BC145586.1 | - | 4 - 5 | 328 | 88 | ||
| BC150797.1 | - | 4 - 5 | 328 | 9087708 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss41
Genomic Map