Assay Details
| Gene Symbol: Prss41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027644.1 | NP_081920.1 | 1 - 2 | 81 | 93 | |
| GenBank mRNA | AB049453.1 | - | 1 - 2 | 81 | 93 | |
| AK049297.1 | - | 1 - 2 | 98 | 93 | ||
| BC145586.1 | - | 1 - 2 | 97 | 93 | ||
| BC150797.1 | - | 1 - 2 | 97 | 9087708 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB049453.1, AK049297.1, BC150797.1, NM_027644.1 transcripts.
Set Membership: |
|

Prss41
Genomic Map