Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700002509.mm.1 |
NM_001080127.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002509.mm.1 |
NM_001080127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002509.mm.1 |
NM_009070.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002509.mm.1 |
NM_009070.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002509.mm.1 |
TC1700000336.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002506.mm.1 |
NM_001080127.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002506.mm.1 |
NM_001080127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002506.mm.1 |
NM_001080128.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002506.mm.1 |
NM_001080128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002506.mm.1 |
NM_009070.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002506.mm.1 |
NM_009070.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002506.mm.1 |
TC1700000336.mm.1 |
MTA TC |