Assay Details
| Gene Symbol: Caskin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027937.2 | NP_082213.2 | 7 - 8 | 919 | 70 | |
| GenBank mRNA | AK014376.1 | - | 7 - 8 | 1096 | 70 | |
| AK129327.1 | - | 7 - 8 | 695 | 70 | ||
| BC060720.1 | - | 7 - 8 | 683 | 70 | ||
| BC138443.1 | - | 7 - 8 | 827 | 8861463 | 70 | |
| BC145281.1 | - | 7 - 8 | 827 | 9053835 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Caskin1
Genomic Map