Assay Details
Gene Symbol: Amdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172935.4 | NP_766523.2 | 6 - 7 | 783 | 78 | |
GenBank mRNA | AK077584.1 | - | 6 - 7 | 766 | 78 | |
AK090009.1 | - | 6 - 7 | 785 | 78 | ||
AK154209.1 | - | 6 - 7 | 784 | 78 | ||
AK159860.1 | - | 6 - 7 | 780 | 78 | ||
AK168432.1 | - | 6 - 7 | 741 | 78 | ||
AK189145.1 | - | 1 - 2 | 66 | 78 | ||
BC037005.1 | - | 6 - 7 | 819 | 5101453 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31095749 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31095748 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31095750 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | AK219479.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | BC037005.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | AK154209.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | AK159860.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | AK168432.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | AK219479.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | BC037005.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | AK154209.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | AK159860.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260658 | AK168432.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635593 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522956 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970472 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299932 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544562 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61629 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114939 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | AK219479.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | BC037005.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | AK154209.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | AK159860.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | AK168432.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | AK219479.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | BC037005.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | AK154209.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | AK159860.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260658 | AK168432.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | AK219479.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | BC037005.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | AK154209.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | AK159860.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | AK168432.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | AK219479.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | BC037005.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | AK154209.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | AK159860.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260658 | AK168432.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095737 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095729 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31095738 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08738618 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114939 | - | - |
Whole Mouse Genome G4122A | A_52_P478460 | AK078259.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | AK219479.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | BC037005.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | AK154209.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | AK159860.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | AK168432.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | AK219479.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | BC037005.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | AK154209.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | AK159860.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260658 | AK168432.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129731 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104743 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114939 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13325239 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31095748 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31095750 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31095749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095812 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095680 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095691 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095700 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095727 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095726 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13078389 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13591282 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095722 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13178134 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13078368 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13078365 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13180948 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13180941 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13078376 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13078366 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095741 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095723 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095682 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095693 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095724 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13180940 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13180956 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095705 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095670 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095717 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095698 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13325247 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095737 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095694 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095684 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095695 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095721 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13325201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095677 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13325234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31095701 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13325239 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10448444 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448454 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448448 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448442 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448451 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448453 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448452 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448450 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448447 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448443 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448449 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434855_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10448441 | - | - |
Mouse Genome 430 2.0 Array | 1434855_at | - | - |
Mouse Genome 430 2.0 Array | 1434855_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245847_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245847_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700014219.mm.1 | NM_172935.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700014219.mm.1 | NM_172935.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700014219.mm.1 | TC1700001618.mm.1 | MTA TC |
Mouse Expression Set 430 | 1434855_at | - | - |
Mouse Expression Set 430 | 1434855_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1434855_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1434855_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 510371 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3610338 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6860338 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006860338 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003610338 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257611 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2605556 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2605556 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257611 | - | - |
mouse-6 v1.1 (Illumina) | scl50218.10.128_2-S | - | - |
mouse-6 v1.1 (Illumina) | scl0001661.1_5-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3610338 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6860338 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001661.1_5-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl50218.10.128_2-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860338 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610338 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2605556 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2831944 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2605556 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2831944 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50218.10.128_2-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001661.1_5-S | - | - |
