| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200013688.mm.1 |
NR_033514.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013688.mm.1 |
NR_033514.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013688.mm.1 |
TC1200001989.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013690.mm.1 |
NR_033514.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013690.mm.1 |
NR_033514.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013690.mm.1 |
TC1200001989.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013684.mm.1 |
NR_033514.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013684.mm.1 |
NR_033514.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013684.mm.1 |
TC1200001989.mm.1 |
MTA TC |