Assay Details
Gene Symbol: Plekhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177503.1 | NP_001170974.1 | 12 - 13 | 1628 | 63 | |
GenBank mRNA | AK028383.1 | - | 12 - 13 | 1578 | 63 | |
AK082841.1 | - | 1 - 2 | 364 | 63 | ||
BC137602.1 | - | 12 - 13 | 1411 | 8860621 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176505 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P157270 | AK028383.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P157270 | NM_001177503.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P157270 | AK028383.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P157270 | NM_001177503.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176505 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02559431 | - | - |
Whole Mouse Genome G4122A | A_51_P157270 | AK028383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P157270 | NM_001177503.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P157270 | AK028383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P157270 | NM_001177503.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176505 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09952167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10207514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10387939 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10222646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577242 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577088 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10303750 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577257 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10317277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09903149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577308 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10303751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577081 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577101 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10317279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577261 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577067 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577070 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09914555 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10272533 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577165 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577258 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577247 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10287533 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577249 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577268 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577103 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577094 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577256 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09952179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577252 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10303757 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09952177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577093 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09896713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09764148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577227 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09896712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577073 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577079 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577236 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577075 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577222 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09903151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10305057 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577269 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577229 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577077 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577262 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10207508 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577310 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10222644 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577260 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09952162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577084 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577090 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577321 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28577298 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10396917 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396911 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396916 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396913 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396906 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396910 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396907 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396915 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396909 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396908 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396912 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396918 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10396914 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436035_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10396905 | - | - |
Mouse Genome 430 2.0 Array | 1436035_at | - | - |
Mouse Genome 430 2.0 Array | 1436035_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217682_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217682_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200005264.mm.1 | NM_001177503.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200005264.mm.1 | NM_001177503.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200005264.mm.1 | TC1200000742.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200005262.mm.1 | NM_001177503.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200005262.mm.1 | NM_001177503.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200005262.mm.1 | TC1200000742.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200005263.mm.1 | NM_001177503.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200005263.mm.1 | NM_001177503.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200005263.mm.1 | TC1200000742.mm.1 | MTA TC |
Mouse Expression Set 430 | 1436035_at | - | - |
Mouse Expression Set 430 | 1436035_PM_at | - | - |
Murine Genome U74v2 Set | 129277_PM_at | - | - |
Murine Genome U74v2 Set | 129277_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436035_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436035_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 762807 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2641486 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2641486 | - | - |
