Assay Details
Gene Symbol: Plekhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177503.1 | NP_001170974.1 | 2 - 3 | 526 | 108 | |
GenBank mRNA | AI747346.1 | - | 2 - 3 | 526 | 108 | |
AK028383.1 | - | 2 - 3 | 476 | 108 | ||
BC137602.1 | - | 2 - 3 | 309 | 8860621 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
