Assay Details
| Gene Symbol: Slc10a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177561.1 | NP_001171032.1 | 2 - 3 | 708 | 63 | |
| NM_011387.2 | NP_035517.1 | 2 - 3 | 708 | 63 | ||
| GenBank mRNA | AB003303.1 | - | 2 - 3 | 633 | 63 | |
| BC021154.1 | - | 2 - 3 | 670 | 4162098 | 63 | |
| BC094023.1 | - | 3 - 4 | 781 | 4160598 | 63 | |
| U95131.1 | - | 2 - 3 | 613 | 63 | ||
| U95132.1 | - | 2 - 3 | 613 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc10a1
Genomic Map