Assay Details
| Gene Symbol: Slc10a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177561.1 | NP_001171032.1 | 4 - 5 | 1076 | 135 | |
| GenBank mRNA | AB003303.1 | - | 4 - 5 | 1001 | 135 | |
| BC021154.1 | - | 4 - 5 | 1038 | 4162098 | 135 | |
| BC094023.1 | - | 5 - 6 | 1149 | 4160598 | 135 | |
| U95131.1 | - | 4 - 5 | 981 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc10a1
Genomic Map