Assay Details
| Gene Symbol: Gm4787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038995.2 | NP_001034084.2 | 1 | 2397 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150248 | BC141094.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150248 | NM_001038995.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406644 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150248 | BC141094.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150248 | NM_001038995.2 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28580998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28580998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09938260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28580971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09938254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28580976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28580951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28580966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28580953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28580958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28580968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28580956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10383426 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10579995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401318 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456368_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10401317 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10579993 | - | - |
| Mouse Genome 430 2.0 Array | 1456368_at | - | - |
| Mouse Genome 430 2.0 Array | 1456368_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 214321_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 214321_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200013927.mm.1 | NM_001038995.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013927.mm.1 | NM_001038995.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013927.mm.1 | TC1200002017.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456368_at | - | - |
| Mouse Expression Set 430 | 1456368_PM_at | - | - |
| Murine Genome U74v2 Set | 166240_PM_r_at | - | - |
| Murine Genome U74v2 Set | 166240_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456368_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456368_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 827094 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160344 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160344 | - | - |
| mouse-6 v1.1 (Illumina) | GI_84781774-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160344 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160344 | - | - |

Gm4787
Genomic Map