Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200014195.mm.1 |
NM_001163501.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014195.mm.1 |
NM_001163501.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014195.mm.1 |
TC1200002070.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200014192.mm.1 |
NM_001163501.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014192.mm.1 |
NM_001163501.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014192.mm.1 |
NM_001163502.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014192.mm.1 |
NM_001163502.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014192.mm.1 |
TC1200002070.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200014193.mm.1 |
NM_001163501.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014193.mm.1 |
NM_001163501.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014193.mm.1 |
NM_001163502.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014193.mm.1 |
NM_001163502.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014193.mm.1 |
TC1200002070.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200014194.mm.1 |
NM_001163502.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014194.mm.1 |
NM_001163502.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014194.mm.1 |
TC1200002070.mm.1 |
MTA TC |