Assay Details
| Gene Symbol: Prox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175198.4 | NP_780407.1 | 1 - 2 | 158 | 89 | |
| GenBank mRNA | AK051700.1 | - | 1 - 2 | 159 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK051700.1, NM_175198.4 transcripts.
Set Membership: |
|

Prox2
Genomic Map