Assay Details
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030225.4 | NP_084501.1 | 11 - 12 | 967 | 102 | |
| GenBank mRNA | AK019713.2 | - | 964 | 102 | ||
| AK054053.1 | - | 11 - 12 | 921 | 102 | ||
| AK149664.1 | - | 11 - 12 | 943 | 102 | ||
| AK158877.1 | - | 11 - 12 | 968 | 102 | ||
| AK168570.1 | - | 11 - 12 | 943 | 102 | ||
| AK169943.1 | - | 11 - 12 | 921 | 102 | ||
| AK197945.1 | - | 2 - 3 | 170 | 102 | ||
| AK220022.1 | - | 2 - 3 | 170 | 102 | ||
| BC006702.1 | - | 11 - 12 | 904 | 3583936 | 102 | |
| BC024066.1 | - | 2 - 3 | 354 | 102 | ||
| CT010197.1 | - | 11 - 12 | 903 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlst
Genomic Map