Assay Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159301.1 | NP_001152773.1 | 8 - 9 | 773 | 251 | |
| NM_010708.2 | NP_034838.2 | 9 - 10 | 866 | 251 | ||
| GenBank mRNA | AK146044.1 | - | 8 - 9 | 729 | 251 | |
| AK214388.1 | - | 3 - 4 | 164 | 251 | ||
| U55060.1 | - | 8 - 9 | 699 | 251 | ||
| U55061.1 | - | 9 - 10 | 773 | 251 | ||
More Information
Set Membership: |
|

Lgals9
Genomic Map