Assay Details
| Gene Symbol: Wsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019653.3 | NP_062627.3 | 2 - 3 | 514 | 122 | |
| GenBank mRNA | AF033186.1 | - | 2 - 3 | 211 | 122 | |
| AK019206.2 | - | 2 - 3 | 491 | 122 | ||
| AK145974.1 | - | 2 - 3 | 493 | 122 | ||
| AK146752.1 | - | 2 - 3 | 489 | 122 | ||
| AK150394.1 | - | 2 - 3 | 489 | 122 | ||
| AK150600.1 | - | 2 - 3 | 489 | 122 | ||
| AK152545.1 | - | 2 - 3 | 492 | 122 | ||
| AK159905.1 | - | 2 - 3 | 489 | 122 | ||
| AK167174.1 | - | 2 - 3 | 491 | 122 | ||
| AK167394.1 | - | 2 - 3 | 492 | 122 | ||
| AK168468.1 | - | 2 - 3 | 492 | 122 | ||
| AK168565.1 | - | 2 - 3 | 492 | 122 | ||
| AK169058.1 | - | 2 - 3 | 489 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wsb1
Genomic Map