Assay Details
| Gene Symbol: Pctp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008796.2 | NP_032822.2 | 3 - 4 | 382 | 69 | |
| GenBank mRNA | AF114437.1 | - | 1 - 2 | 87 | 69 | |
| AF151639.1 | - | 3 - 4 | 69 | |||
| BB614866.1 | - | 230 | 69 | |||
| BC053478.1 | - | 337 | 69 | |||
| BC071234.1 | - | 3 - 4 | 338 | 69 | ||
| Z50024.1 | - | 3 - 4 | 211 | 69 | ||
More Information
Set Membership: |
|

Pctp
Genomic Map