Assay Details
| Gene Symbol: Akap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104310.1 | 3 - 4 | 1874 | 75 | ||
| GenBank mRNA | AK186415.1 | - | 2 - 3 | 76 | ||
| U95145.1 | - | 3 - 4 | 1878 | 75 | ||
More Information
Set Membership: |
|

Akap1
Genomic Map