Assay Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284392.1 | NP_001271321.1 | 9 - 10 | 1081 | 62 | |
| NM_011129.2 | NP_035259.1 | 10 - 11 | 1378 | 62 | ||
| GenBank mRNA | AB078010.1 | - | 9 - 10 | 976 | 62 | |
| AK135143.1 | - | 10 - 11 | 1379 | 62 | ||
| AW123821.1 | - | 2 - 3 | 62 | |||
| X61452.1 | - | 10 - 11 | 1308 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sept4
Genomic Map