Assay Details
| Gene Symbol: Pcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103166.1 | NP_001096636.1 | 8 - 9 | 824 | 65 | |
| NM_001174073.1 | NP_001167544.1 | 8 - 9 | 812 | 65 | ||
| GenBank mRNA | AK161527.1 | - | 8 - 9 | 660 | 65 | |
| AK213763.1 | - | 1 - 2 | 45 | 65 | ||
| BC020478.1 | - | 7 - 8 | 569 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcbp2
Genomic Map