Assay Details
Gene Symbol: Mtch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019880.3 | NP_063933.1 | 7 - 8 | 880 | 102 | |
GenBank mRNA | AF176007.2 | - | 7 - 8 | 881 | 102 | |
AF192558.2 | - | 7 - 8 | 881 | 102 | ||
AK154010.1 | - | 7 - 8 | 736 | 102 | ||
AK178745.1 | - | 2 - 3 | 116 | 102 | ||
AK183124.1 | - | 2 - 3 | 116 | 102 | ||
AK195241.1 | - | 2 - 3 | 116 | 102 | ||
AK199429.1 | - | 2 - 3 | 116 | 102 | ||
AK200271.1 | - | 2 - 3 | 117 | 102 | ||
AK201694.1 | - | 2 - 3 | 116 | 102 | ||
AK207873.1 | - | 2 - 3 | 116 | 102 | ||
BC023135.1 | - | 7 - 8 | 723 | 3669288 | 102 | |
BC026436.1 | - | 3 - 4 | 165 | 102 | ||
BC027274.1 | - | 7 - 8 | 729 | 3156729 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
