Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500018170.mm.1 |
NM_001253808.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018170.mm.1 |
NM_001253808.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018170.mm.1 |
NM_001253809.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018170.mm.1 |
NM_001253809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018170.mm.1 |
NM_012025.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018170.mm.1 |
NM_012025.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018170.mm.1 |
TC1500002217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018168.mm.1 |
NM_001253808.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018168.mm.1 |
NM_001253808.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018168.mm.1 |
NM_001253809.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018168.mm.1 |
NM_001253809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018168.mm.1 |
NM_012025.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018168.mm.1 |
NM_012025.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018168.mm.1 |
TC1500002217.mm.1 |
MTA TC |