Assay Details
| Gene Symbol: Stard3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021547.3 | NP_067522.1 | 9 - 10 | 991 | 72 | |
| GenBank mRNA | AK041538.1 | - | 9 - 10 | 947 | 72 | |
| AK150166.1 | - | 9 - 10 | 935 | 72 | ||
| AK150254.1 | - | 9 - 10 | 935 | 72 | ||
| AK155124.1 | - | 9 - 10 | 930 | 72 | ||
| AK160480.1 | - | 9 - 10 | 911 | 72 | ||
| AK197418.1 | - | 1 - 2 | 90 | 72 | ||
| BC003313.1 | - | 9 - 10 | 908 | 3155179 | 72 | |
| BC058727.1 | - | 10 - 11 | 1078 | 72 | ||
| X82457.1 | - | 9 - 10 | 975 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard3
Genomic Map