Assay Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013871.3 | NP_038899.1 | 9 - 10 | 1003 | 104 | |
| GenBank mRNA | AK011286.1 | - | 8 - 9 | 1043 | 104 | |
| BC021640.1 | - | 9 - 10 | 921 | 3979667 | 104 | |
| Y13439.1 | - | 9 - 10 | 939 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk12
Genomic Map